CH6371384410 Chem Impex International, Inc.

3,5-Dinitro-L-tyrosine monohydrate

  • MDL number MFCD00150534
  • Cas Number : 17360-11-1
  • Molecular Weight : 289.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Store at 0-8 °C
Physical Form Bright red powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/3-5-dinitro-l-tyrosine-monohydrate/11966?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/3-5-dinitro-l-tyrosine-monohydrate/11966?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart