CH6371384412 Chem Impex International, Inc.

4-Methylumbelliferyl elaidate

  • MDL number MFCD00037610
  • Cas Number : 69003-01-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Store at 0-8 °C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/4-methylumbelliferyl-elaidate/11969?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/4-methylumbelliferyl-elaidate/11969?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart