CH6371384423 Chem Impex International, Inc.

5-Ethylindole

  • MDL number MFCD00047201
  • Cas Number : 68742-28-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Store at 0-8 °C
Physical Form Colorless to yellow liquid
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/5-ethylindole/11983?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/5-ethylindole/11983?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart