CH6371384429 Chem Impex International, Inc.

6-Bromo-DL-tryptophan

  • MDL number MFCD00055988
  • Cas Number : 33599-61-0
  • Molecular Weight : 283.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Store at 0-8 °C
Physical Form White to pale yellow powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/6-bromo-dl-tryptophan/11992?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/6-bromo-dl-tryptophan/11992?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart