CH6371384430 Chem Impex International, Inc.

6-Chloro-3-indolyl phosphate p-toluidine salt

  • MDL number MFCD00216690
  • Cas Number : 159954-33-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Store at 0-8°C
Physical Form White to off-white crystalline powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/6-chloro-3-indolyl-phosphate-i-p-i-toluidine-salt/11993?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/6-chloro-3-indolyl-phosphate-i-p-i-toluidine-salt/11993?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart