CH6371384435 Chem Impex International, Inc.

7-Methylindole-3-acetic acid

  • MDL number MFCD00047183
  • Cas Number : 5435-36-9
  • Molecular Weight : 189.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 178-186 °C
Purity 99
Storage temp. Store at 0-8 °C
Physical Form Yellowish to off-white crystalline powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/7-methylindole-3-acetic-acid/11999?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/7-methylindole-3-acetic-acid/11999?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart