CH6371384439 Chem Impex International, Inc.

Sodium tauroglycocholate

  • MDL number MFCD00036243
  • Cas Number : 11006-55-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Storage temp. Store at 0-8 °C
Physical Form White to yellow crystalline powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/sodium-tauroglycocholate/12005?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/sodium-tauroglycocholate/12005?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart