CH6371384446 Chem Impex International, Inc.

2,3,4,6-Tetra-O-benzyl-D-mannopyranose

  • MDL number MFCD00153594
  • Cas Number : 61330-61-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Store at 0-8 °C
Physical Form Colorless to pale yellow oil
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/2-3-4-6-tetra-o-benzyl-d-mannopyranose/12013?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/2-3-4-6-tetra-o-benzyl-d-mannopyranose/12013?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart