CH6371384452 Chem Impex International, Inc.

2-Methyl-DL-tryptophan

  • MDL number MFCD03005300
  • Cas Number : 21495-41-0
  • Molecular Weight : 218.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Store at 0-8 °C
Physical Form Off-white solid
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/2-methyl-dl-tryptophan/12020?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/2-methyl-dl-tryptophan/12020?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart