CH6371384460 Chem Impex International, Inc.

3-Nitro-D-tyrosine

  • MDL number MFCD02684446
  • Cas Number : 32988-39-9
  • Molecular Weight : 226.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point > 218 °C (dec.)
Purity 98
Storage temp. Store at 0-8 °C
Physical Form Yellow powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/3-nitro-d-tyrosine/12033?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/3-nitro-d-tyrosine/12033?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart