CH6371384466 Chem Impex International, Inc.

4-Methylumbelliferyl propionate

  • MDL number MFCD00047644
  • Cas Number : 3361-13-5
  • Molecular Weight : 232.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Storage temp. Store at 0 - 8 °C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/4-methylumbelliferyl-propionate/12042?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/4-methylumbelliferyl-propionate/12042?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart