CH6371384468 Chem Impex International, Inc.

4-Methylumbelliferyl-beta-D(-)-glucuronide dihydrate

  • MDL number MFCD00151678
  • Cas Number : 6160-80-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Storage temp. Store at 0-8 °C
Physical Form White crystalline powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/4-methylumbelliferyl-beta-d-glucuronide-dihydrate/12044?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/4-methylumbelliferyl-beta-d-glucuronide-dihydrate/12044?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart