CH6371384486 Chem Impex International, Inc.

5-Bromo-4-chloro-3-indoxyl-alpha-L-fucopyranoside

  • MDL number MFCD00132991
  • Cas Number : 171869-92-4
  • Molecular Weight : 392.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Storage temp. Store at 0-8°C
Physical Form White to slightly off-white powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/5-bromo-4-chloro-3-indoxyl-alpha-l-fucopyranoside/12065?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/5-bromo-4-chloro-3-indoxyl-alpha-l-fucopyranoside/12065?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart