CH6371384491 Chem Impex International, Inc.

5-Bromo-6-chloro-3-indolyl caprylate

  • MDL number MFCD00798386
  • Cas Number : 209347-94-4
  • Molecular Weight : 372.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 58-63 °C
Purity 98
Storage temp. Store at 0-8 °C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/5-bromo-6-chloro-3-indolyl-caprylate/12070?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/5-bromo-6-chloro-3-indolyl-caprylate/12070?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart