CH6371384500 Chem Impex International, Inc.

5-Ethoxy-1H-indole-3-carbaldehyde

  • MDL number MFCD07364715
  • Cas Number : 169789-47-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 5-Ethoxy-3-indolecarboxaldehyde
Purity 95
Storage temp. Store at 0-8
Physical Form Yellow to pale brown solid
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/5-ethoxy-1-i-h-i-indole-3-carbaldehyde/12079?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/5-ethoxy-1-i-h-i-indole-3-carbaldehyde/12079?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart