CH6371384557 Chem Impex International, Inc.

4-Methylumbelliferyl pyrophosphate diester disodium salt

  • MDL number MFCD00051219
  • Cas Number : 84282-11-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Storage temp. Store at 0-8°C
Physical Form White to off-white crystalline powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/4-methylumbelliferyl-pyrophosphate-diester-disodium-salt/12203?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/4-methylumbelliferyl-pyrophosphate-diester-disodium-salt/12203?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart