CH6371384612 Chem Impex International, Inc.

2-Naphthyl-beta-D-galactopyranoside

  • MDL number MFCD00004056
  • Cas Number : 33993-25-8
  • Molecular Weight : 306.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Store at 0-8°C
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/2-naphthyl-ß-d-galactopyranoside/12263?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/2-naphthyl-ß-d-galactopyranoside/12263?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart