CH6371384708 Chem Impex International, Inc.

5-Bromoindole-3-acetamide

  • MDL number MFCD00022752
  • Cas Number : 196081-79-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 178-182 °C
Purity 98
Storage temp. Store at 0-8°C
Physical Form White to off-white crystalline powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/5-bromoindole-3-acetamide/12367?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/5-bromoindole-3-acetamide/12367?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart