CH6371384709 Chem Impex International, Inc.

5-Chloroindole-2-carboxylic acid methyl ester

  • MDL number MFCD04972050
  • Cas Number : 87802-11-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 215-219 °C
Purity 99
Storage temp. Store at 0-8°C
Physical Form Slightly off-white crystalline powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/5-chloroindole-2-carboxylic-acid-methyl-ester/12368?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/5-chloroindole-2-carboxylic-acid-methyl-ester/12368?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart