CH6371384714 Chem Impex International, Inc.

5-Methylindole-3-acetic acid

  • MDL number MFCD00022751
  • Cas Number : 1912-47-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 150-158 �C
Synonym (5-Methyl-1H-indol-3-yl)acetic acid
Purity 99
Storage temp. Store at 0-8
Physical Form Off-white crystalline powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/5-methylindole-3-acetic-acid/12374?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/5-methylindole-3-acetic-acid/12374?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart