CH6371384721 Chem Impex International, Inc.

6-Chloroindole-3-acetonitrile

  • MDL number MFCD03428598
  • Cas Number : 61220-58-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 101 - 107 °C
Purity 98
Storage temp. Store at 0-8°C
Physical Form Light yellow crystal
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/6-chloroindole-3-acetonitrile/12381?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/6-chloroindole-3-acetonitrile/12381?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart