CH6371384725 Chem Impex International, Inc.

7-Methoxytryptamine

  • MDL number MFCD01115208
  • Cas Number : 2436-04-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 135-139 °C
Purity 99
Storage temp. Store at 0-8 °C
Physical Form White to off-white crystalline powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/7-methoxytryptamine/12385?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/7-methoxytryptamine/12385?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart