CH6371384774 Chem Impex International, Inc.

5-Bromo-6-chloro-3-indoxyl-beta-D-glucopyranoside

  • MDL number MFCD00210022
  • Cas Number : 93863-89-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Storage temp. Store at 0-8 °C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/5-bromo-6-chloro-3-indoxyl-ß-d-glucopyranoside/12444?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/5-bromo-6-chloro-3-indoxyl-ß-d-glucopyranoside/12444?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart