CH6371384777 Chem Impex International, Inc.

5-Chlorogramine

  • MDL number MFCD01074497
  • Cas Number : 830-94-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Store at 0-8 °C
Physical Form White to off-white crystalline powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/5-chlorogramine/12447?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/5-chlorogramine/12447?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart