CH6371384784 Chem Impex International, Inc.

5-Methylgramine

  • MDL number MFCD00022732
  • Cas Number : 30218-58-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 3-(Dimethylaminomethyl)-5-methylindole
Purity 98
Storage temp. Store at 0-8
Physical Form White to off-white crystalline powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/5-methylgramine/12457?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/5-methylgramine/12457?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart