CH6371384809 Chem Impex International, Inc.

L-Tryptophan-7-amido-4-methylcoumarin dihydrochloride

  • MDL number MFCD02683915
  • Cas Number : 201860-49-3
  • Molecular Weight : 397.86

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Storage temp. Store at 0-8°C
Physical Form Pale violet powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/l-tryptophan-7-amido-4-methylcoumarin-dihydrochloride/12502?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/l-tryptophan-7-amido-4-methylcoumarin-dihydrochloride/12502?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart