CH6371384819 Chem Impex International, Inc.

Phosphoenolpyruvic acid trisodium salt heptahydrate

  • MDL number MFCD00150737
  • Cas Number : 5541-93-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Storage temp. Store at 0-8 °C
Physical Form White to off-white crystalline powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/phosphoenolpyruvic-acid-trisodium-salt-heptahydrate/12515?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/phosphoenolpyruvic-acid-trisodium-salt-heptahydrate/12515?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart