CH6371384835 Chem Impex International, Inc.

5-Fluorogramine

  • MDL number MFCD00022731
  • Cas Number : 343-90-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym (5-Fluoro-1H-indol-3-yl-methyl)dimethylamine
Purity 99
Storage temp. Store at 0-8
Physical Form White to off-white crystalline powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/5-fluorogramine/12542?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/5-fluorogramine/12542?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart