CH6371384858 Chem Impex International, Inc.

6-Fluoroindole-3-acetic acid

  • MDL number MFCD01074503
  • Cas Number : 443-75-4
  • Molecular Weight : 193.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Store at 0-8 °C
Physical Form Brown powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/6-fluoroindole-3-acetic-acid/12570?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/6-fluoroindole-3-acetic-acid/12570?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart