CH6371384859 Chem Impex International, Inc.

5-Chloroindole-3-carboxylic acid

  • MDL number MFCD03410308
  • Cas Number : 10406-05-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 5-Chloro-1H-indole-3-carboxylic acid
Purity 98
Storage temp. Store at 0-8
Physical Form Off-white powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/5-chloroindole-3-carboxylic-acid/12571?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/5-chloroindole-3-carboxylic-acid/12571?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart