CH6371384872 Chem Impex International, Inc.

Muristerone A

  • MDL number MFCD00056450
  • Cas Number : 38778-30-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Store at 0-8°C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/muristerone-a/12585?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/muristerone-a/12585?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart