CH6371384903 Chem Impex International, Inc.

Methyl indole-2-carboxylate

  • MDL number MFCD00460779
  • Cas Number : 1202-04-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1H-Indole-2-carboxylic acid methyl ester
Purity 98
Storage temp. Store at 0-8
Physical Form Off-white to pinkish powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/methyl-indole-2-carboxylate/12616?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/methyl-indole-2-carboxylate/12616?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart