CH6371384920 Chem Impex International, Inc.

Ethyl 5-hydroxyindole-2-carboxylate

  • MDL number MFCD00015458
  • Cas Number : 24985-85-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 150-155 �C
Synonym 2-Carbethoxy-5-hydroxyindole
Purity 98
Storage temp. Store at 0-8
Physical Form Almost white to pale yellow powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/ethyl-5-hydroxyindole-2-carboxylate/12633?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/ethyl-5-hydroxyindole-2-carboxylate/12633?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart