CH6371385009 Chem Impex International, Inc.

6-Fluorotryptamine

  • MDL number MFCD00044761
  • Cas Number : 575-85-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 80-88 °C
Purity 97
Storage temp. Store at = -4 °C
Physical Form Yellow crystalline powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/6-fluorotryptamine/12732?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/6-fluorotryptamine/12732?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart