CH6371385011 Chem Impex International, Inc.

Gentiobiose

  • MDL number MFCD00198056
  • Cas Number : 554-91-6
  • Molecular Weight : 342.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 180 - 190
Storage temp. Store at 0 - 8
Physical Form White to off-white powder
Shipping Temperature RT
Country of Origin US
Product Link http://www.chemimpex.com/gentiobiose

Safety Information

MSDS http://www.chemimpex.com/gentiobiose
Availability details for
Quantity
  • Availability
  • Add to Cart