CH6371385024 Chem Impex International, Inc.

5-Bromo-4-chloro-3-indolyl-N-acetyl-beta-D-glucosaminide

  • MDL number MFCD00040631
  • Cas Number : 4264-82-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Storage temp. Store at 0-8 °C
Physical Form White to off-white crystalline powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/5-bromo-4-chloro-3-indolyl-n-acetyl-beta-d-glucosaminide/12748?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/5-bromo-4-chloro-3-indolyl-n-acetyl-beta-d-glucosaminide/12748?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart