CH6371385057 Chem Impex International, Inc.

G-D-Galactonolactone

  • MDL number MFCD00065019
  • Cas Number : 2782-07-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Store at 0-8°C
Physical Form White to off-white solid
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/d-galactonolactone/12782?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/d-galactonolactone/12782?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart