CH6371385080 Chem Impex International, Inc.

1,2:3,5-Di-o-isopropylidene-alpha-D-xylofuranose

  • MDL number MFCD00063224
  • Cas Number : 20881-04-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 38-47 °C
Purity 99
Storage temp. Store at 0-8 °C
Physical Form White crystalline powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/1-2-3-5-di-o-isopropylidene-a-d-xylofuranose/12807?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/1-2-3-5-di-o-isopropylidene-a-d-xylofuranose/12807?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart