CH6371385116 Chem Impex International, Inc.

N-Acetyl-3,5-diiodo-L-tyrosine

  • MDL number MFCD00020004
  • Cas Number : 1027-28-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 3,5-Diiodo-N-acetyl-L-tyrosine
Purity 97
Storage temp. Store at 0-8
Physical Form Light yellow crystalline powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-acetyl-3-5-diiodo-l-tyrosine/12852?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-acetyl-3-5-diiodo-l-tyrosine/12852?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart