CH6371385118 Chem Impex International, Inc.

Bis(4-nitrophenyl)phosphoric acid sodium salt , [for Phosphodiesterase substrate]

  • MDL number MFCD00065378
  • Cas Number : 4043-96-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Storage temp. Store at 0-8°C
Physical Form White to light yellow powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/bis-4-nitrophenyl-phosphoric-acid-sodium-salt/12854?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/bis-4-nitrophenyl-phosphoric-acid-sodium-salt/12854?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart