CH6371385119 Chem Impex International, Inc.

5-Hydroxyoxindole

  • MDL number MFCD00014566
  • Cas Number : 3416-18-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Store at RT
Physical Form Off-white to yellow powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/5-hydroxyoxindole/12855?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/5-hydroxyoxindole/12855?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart