CH6371385410 Chem Impex International, Inc.

N,N'-Di(n-propyl)ethylenediamine

  • MDL number MFCD00015199
  • Cas Number : 14165-22-1
  • Molecular Weight : 144.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Density 0.802 g/mL at 25
Flash Point 69 °C
Storage temp. Store at 0-8 °C
Physical Form Clear light yellow liquid
Shipping Temperature RT
Country of Origin USA
Boiling point 186-189 °C
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-i-n-i-di-n-propyl-ethylenediamine/13150?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-i-n-i-di-n-propyl-ethylenediamine/13150?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart