CH6371385412 Chem Impex International, Inc.

5-Phenylpentyl chloride

  • MDL number MFCD00037118
  • Cas Number : 15733-63-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Store at 0-8°C
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/5-phenylpentyl-chloride/13154?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/5-phenylpentyl-chloride/13154?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart