CH6371385429 Chem Impex International, Inc.

3-Ethylbenzonitrile

  • MDL number MFCD00045601
  • Cas Number : 34136-57-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Store at 0-8
Physical Form Colorless to brown liquid
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/3-ethylbenzonitrile/13174?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/3-ethylbenzonitrile/13174?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart