CH6371385440 Chem Impex International, Inc.

9-Heptadecanone

  • MDL number MFCD00009573
  • Cas Number : 540-08-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 51-53 °C
Purity 98
Storage temp. Store at RT
Physical Form White to off-white crystalline powder
Shipping Temperature RT
Country of Origin USA
Boiling point 250-253 °C
Product Link http://www.chemimpex.com/product/productinfo/di-n-octylketone/13191?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/di-n-octylketone/13191?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart