CH6371385459 Chem Impex International, Inc.

Fmoc-threo-beta-phenylserine

  • MDL number MFCD06656479
  • Cas Number : 487060-72-0
  • Molecular Weight : 403.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 150-154 °C
Purity 99
Storage temp. Store at 0-8 °C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-i-threo-i-beta-phenylserine/13211?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-i-threo-i-beta-phenylserine/13211?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart