CH6371385462 Chem Impex International, Inc.

Boc-methylaminooxyacetic acid�A

  • MDL number MFCD06656434
  • Cas Number : 1185005-62-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 119-128 °C
Purity 98
Storage temp. Store at 0-8 °C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-methylaminooxyacetic-acid-font-size-2-face-symbol-middot-font-dcha/13214?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-methylaminooxyacetic-acid-font-size-2-face-symbol-middot-font-dcha/13214?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart