CH6371385466 Chem Impex International, Inc.

N-Fmoc-erythro-DL-beta-methylphenylalanine

  • MDL number MFCD06656477
  • Cas Number : 321524-79-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 151-156 °C
Purity 99
Storage temp. Store at 0-8 °C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-fmoc-i-erythro-i-dl-beta-methylphenylalanine/13218?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-fmoc-i-erythro-i-dl-beta-methylphenylalanine/13218?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart