CH6371385468 Chem Impex International, Inc.

Fmoc-N-methyl-tranexamic acid

  • MDL number MFCD06656462
  • Cas Number : 173690-50-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98.5
Storage temp. Store at 0-8 °C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-i-n-i-methyl-tranexamic-acid/13220?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-i-n-i-methyl-tranexamic-acid/13220?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart