CH6371385469 Chem Impex International, Inc.

(2S,4R)-Boc-4-(2-naphthylmethoxy)pyrrolidine-2-carboxylic acid

  • MDL number MFCD06656451
  • Cas Number : 630425-40-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Storage temp. Store at 0-8 °C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/2-i-s-i-4-i-r-i-boc-4-2-naphthylmethoxy-pyrrolidine-2-carboxylic-acid/13221?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/2-i-s-i-4-i-r-i-boc-4-2-naphthylmethoxy-pyrrolidine-2-carboxylic-acid/13221?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart